Learn More
Waters Corp Symmetry Prep Guard Cartridge, C8, 7 μm, 19 mm x 10 mm
Catalog No. 50780481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry Prep Guard Cartridges set the standard for HPLC drug assays, ensuring long-term compliance for HPLC methods. They maintain excellent peak shape, selectivity, and reproducibility, offering consistent performance across columns and batches.
  • Sets the standard for HPLC drug assays.
  • Provides confidence in long-term compliance of HPLC methods.
  • Maintains peak shape, selectivity, and reproducibility.
  • Available in various particle sizes, chemistries, and dimensions.
Catalog No. 50-780-481 Supplier Waters Corp Supplier No. 186000718
Symmetry Prep Guard Cartridges set the standard for HPLC drug assays, ensuring long-term compliance for HPLC methods. They maintain excellent peak shape, selectivity, and reproducibility, offering consistent performance across columns and batches.
  • Sets the standard for HPLC drug assays.
  • Provides confidence in long-term compliance of HPLC methods.
  • Maintains peak shape, selectivity, and reproducibility.
  • Available in various particle sizes, chemistries, and dimensions.