Learn More
Waters Corp Waters Corporation Symmetry Prep Column, C8, 5 μm, 19 mm × 100 mm
Catalog No. 50780140 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry columns are designed to set the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods. They maintain peak shape, selectivity, column-to-column, and batch-to-batch reproducibility, along with other critical performance characteristics. These columns are available in various particle sizes, chemistries, and dimensions.
  • Optimized for HPLC drug assays.
  • Ensures long-term compliance of HPLC methods.
  • Maintains peak shape and selectivity.
  • Offers column-to-column and batch-to-batch reproducibility.
  • Available in a variety of particle sizes, chemistries, and dimensions.
Catalog No. 50-780-140 Supplier Waters Corp Supplier No. 186000229
Symmetry columns are designed to set the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods. They maintain peak shape, selectivity, column-to-column, and batch-to-batch reproducibility, along with other critical performance characteristics. These columns are available in various particle sizes, chemistries, and dimensions.
  • Optimized for HPLC drug assays.
  • Ensures long-term compliance of HPLC methods.
  • Maintains peak shape and selectivity.
  • Offers column-to-column and batch-to-batch reproducibility.
  • Available in a variety of particle sizes, chemistries, and dimensions.