Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Symmetry Preparative Column, C8, 5 μm, 7.8 mm x 100 mm

Catalog No. 50780134 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Symmetry preparative columns are optimized for HPLC drug assays, ensuring reliable and compliant analytical methods. They provide consistent peak shape, excellent selectivity, and dependable reproducibility across columns and batches, available in various particle sizes, chemistries, and dimensions.

  • Ensures consistent peak shape, excellent selectivity, and dependable reproducibility
  • Optimized for HPLC drug assays and long-term method compliance
  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped silica particles
  • Spherical particle shape
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Dimensions: 7.8 mm inner diameter × 100 mm length

Catalog No. 50-780-134 Supplier Waters Corp Supplier No. 186000215
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Symmetry preparative columns are optimized for HPLC drug assays, ensuring reliable and compliant analytical methods. They provide consistent peak shape, excellent selectivity, and dependable reproducibility across columns and batches, available in various particle sizes, chemistries, and dimensions.

  • Ensures consistent peak shape, excellent selectivity, and dependable reproducibility
  • Optimized for HPLC drug assays and long-term method compliance
  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped silica particles
  • Spherical particle shape
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Dimensions: 7.8 mm inner diameter × 100 mm length

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.