Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Prep Column, C8, 5 μm, 100 Å, 30 mm × 100 mm

Supplier:  Waters Corp 186000238

Encompass_Preferred

Symmetry columns set the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods. They provide consistent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility. These columns are available in various particle sizes, chemistries, and dimensions.

  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range: 2 to 8 pH
  • Temperature limits: 45 °C
  • Maximum pressure: 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • Particle size: 5 μm
  • Standard endfitting type
  • Pore size: 100 Å
  • Surface area: 335
  • System: HPLC preparative
  • Techniques: LC/MS, LC
  • USP classification: L7
  • Inner diameter: 30 mm
  • Length: 100 mm
  • Carbon load: 12%
  • 1 unit per package

Catalog No. 50-780-145


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.