Learn More
Waters Corp Symmetry Shield RP18 Sentry Guard Cartridge, 100Å, 3.5 µm, 2.1 mm X 10 mm, 2/pkg
Catalog No. 50819047 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP18 column significantly improves peak shape and resolution.
Catalog No. 50-819-047 Supplier Waters Corp Supplier No. 186000169
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP18 column significantly improves peak shape and resolution.