Learn More
Waters Corp Waters Symmetry Shield RP18 Prep Guard Cartridge, 100Å, 5 μm
Catalog No. 50780765 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This reversed-phase Prep Guard Cartridge incorporates embedded polar group technology, which shields the silica's residual silanol surface from highly basic analytes. The reduced silanol activity significantly improves peak shape and resolution. This product is designed to be used as part of a preparative HPLC workflow. A Prep Guard Cartridge serves to protect analytical or preparative columns by removing contaminants from samples.
  • Embedded polar group technology shields the silica's residual silanol surface from highly basic analytes.
  • Improved peak shape and resolution is achieved through reduced silanol activity.
  • Protects analytical or preparative columns by removing contaminants from samples.
  • Reduced silanol activity is important for LC-MS analysis.
  • Narrow column specification reduces data variation.
  • Can be used with peptide applications.
Catalog No. 50-780-765 Supplier Waters Corp Supplier No. 186001835
This reversed-phase Prep Guard Cartridge incorporates embedded polar group technology, which shields the silica's residual silanol surface from highly basic analytes. The reduced silanol activity significantly improves peak shape and resolution. This product is designed to be used as part of a preparative HPLC workflow. A Prep Guard Cartridge serves to protect analytical or preparative columns by removing contaminants from samples.
  • Embedded polar group technology shields the silica's residual silanol surface from highly basic analytes.
  • Improved peak shape and resolution is achieved through reduced silanol activity.
  • Protects analytical or preparative columns by removing contaminants from samples.
  • Reduced silanol activity is important for LC-MS analysis.
  • Narrow column specification reduces data variation.
  • Can be used with peptide applications.