Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, SymmetryShield RP18, 3.5 μm, 2.1 x 30 mm

Supplier:  Waters Corp 186000171

Encompass_Preferred

This Waters SymmetryShield RP18 column is designed for use in chromatography applications.

  • Used in chromatography.
  • Features a 3.5 μm particle size.
  • Has dimensions of 2.1 x 30 mm.

Catalog No. 50-780-099


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.