Learn More
Waters Corp Waters Corporation HPLC Column, 100Å, 3.5 μm, 2.1 mm x 150 mm
Catalog No. 50780102 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns incorporate patented embedded polar group technology, designed to “shield” the silica’s residual silanol surface from highly basic analytes. This significantly enhances peak shape and resolution by reducing silanol activity. The column offers outstanding reproducibility due to narrow specification ranges, providing the industry’s narrowest ligand surface coverage and minimizing resolution shifts batch-to-batch and column-to-column.
  • Enhances peak shape and resolution with reduced silanol activity.
  • Ensures high reproducibility and minimal shifts between batches and columns.
  • Durable design with hydrolytic and mechanical stability for extended lifetime.
  • Operates effectively across a 2 to 8 pH range, supporting method optimization.
  • Suitable for HPLC applications, offering lower detection and quantitation levels.
Catalog No. 50-780-102 Supplier Waters Corp Supplier No. 186000174
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These reversed-phase columns incorporate patented embedded polar group technology, designed to “shield” the silica’s residual silanol surface from highly basic analytes. This significantly enhances peak shape and resolution by reducing silanol activity. The column offers outstanding reproducibility due to narrow specification ranges, providing the industry’s narrowest ligand surface coverage and minimizing resolution shifts batch-to-batch and column-to-column.
  • Enhances peak shape and resolution with reduced silanol activity.
  • Ensures high reproducibility and minimal shifts between batches and columns.
  • Durable design with hydrolytic and mechanical stability for extended lifetime.
  • Operates effectively across a 2 to 8 pH range, supporting method optimization.
  • Suitable for HPLC applications, offering lower detection and quantitation levels.