Learn More
Waters Corp Waters Column, Symmetry Shield RP18, 3.5 μm, 4.6 mm, 75 mm
Catalog No. 50780106 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns feature patented embedded polar group technology, which “shields” the residual silanol surface of the silica from highly basic analytes. The unique integrated polar group technology and its closeness to the silica surface effectively protect leftover surface silanols, reducing silanol activity for significantly improved peak shape and resolution.
  • Ideal for aqueous mobile phases, requiring low organic conditions.
  • Prevents pore dewetting by preserving the wetted-pore surface.
  • Ensures analyte retention, eliminating lengthy re-wetting and column re-equilibration.
  • Provides small ligand surface coverage for exceptional reproducibility.
  • Improves batch-to-batch and column-to-column repeatability.
  • Offers extended durability through hydrolytic and mechanical stability, saving lab equipment costs.
Catalog No. 50-780-106 Supplier Waters Corp Supplier No. 186000178
These reversed-phase columns feature patented embedded polar group technology, which “shields” the residual silanol surface of the silica from highly basic analytes. The unique integrated polar group technology and its closeness to the silica surface effectively protect leftover surface silanols, reducing silanol activity for significantly improved peak shape and resolution.
  • Ideal for aqueous mobile phases, requiring low organic conditions.
  • Prevents pore dewetting by preserving the wetted-pore surface.
  • Ensures analyte retention, eliminating lengthy re-wetting and column re-equilibration.
  • Provides small ligand surface coverage for exceptional reproducibility.
  • Improves batch-to-batch and column-to-column repeatability.
  • Offers extended durability through hydrolytic and mechanical stability, saving lab equipment costs.