Learn More
Waters Corp SymmetryShield RP18 Column, 5 μm/100Å, 19 x 100 mm
Supplier: Waters Corp 186001837
Symmetry columns are designed for HPLC drug assays, offering consistent peak shape, selectivity, and reproducibility across columns and batches. These columns are available in various particle sizes, chemistries, and dimensions.
- Carbon load: 17%
- Chemistry: C18
- Endcapped: Yes
- Endfitting type: Proprietary design
- Format: Column
- Inner diameter: 19 mm
- Length: 100 mm
- Mode: Reversed-phase
- Particle shape: Spherical
- Particle size: 5 μm
- Particle substrate: Silica
- Pore size: 100Å
- Silanol activity: Low
- Surface area: 335
- USP classification: L1
- Units in package: 1/pkg
- pH range: 2 to 8
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.