Learn More
Waters Corp Waters Column, C18, 5 μm, 4.6 mm, 50 mm
Supplier:  Waters Corp 186000218
Encompass_Preferred
These reversed-phase columns utilize patented embedded polar group technology to “shield” the silica’s residual silanol surface from highly basic analytes. This leads to improved peak shape and resolution. Designed for consistent chromatographic findings, these columns are ideal for developing new analytical techniques for pharmaceutical and biopharmaceutical products.
  • Reduced silanol activity enhances peak shape and resolution for basic analytes.
  • Embedded polar group technology protects the silica surface, allowing superior peak form across a wide pH range.
  • Delivers consistent and repeatable chromatographic results.
  • Decreased retention for basic chemicals aids method development.
  • Allows operation at neutral or low pH levels without compromising peak shape.
  • Suitable for developing new analytical techniques in pharmaceutical and biopharmaceutical fields.
Catalog No. 50-780-137