Learn More
Waters Corp Waters Column, 5 μm, 100 Å, 3.9 mm x 150 mm
Catalog No. 50780068 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns utilize patented embedded polar group technology to shield the silica's residual silanol surface from highly basic analytes. This results in significantly improved peak shape and resolution, particularly for basic compounds, and offers consistent performance across a wide pH range.
  • Contains patented embedded polar group technology for improved performance with basic analytes.
  • Provides significantly improved peak shape and resolution by reducing silanol activity.
  • Ensures exceptional reproducibility through narrow specification ranges and minimal ligand surface coverage.
  • Designed for longevity, with hydrolytically stable packing material and a mechanically stable packed bed.
  • Supports a wide pH range (2–8), allowing method optimization without compromising peak shape.
Catalog No. 50-780-068 Supplier Waters Corp Supplier No. 186000108
These reversed-phase columns utilize patented embedded polar group technology to shield the silica's residual silanol surface from highly basic analytes. This results in significantly improved peak shape and resolution, particularly for basic compounds, and offers consistent performance across a wide pH range.
  • Contains patented embedded polar group technology for improved performance with basic analytes.
  • Provides significantly improved peak shape and resolution by reducing silanol activity.
  • Ensures exceptional reproducibility through narrow specification ranges and minimal ligand surface coverage.
  • Designed for longevity, with hydrolytically stable packing material and a mechanically stable packed bed.
  • Supports a wide pH range (2–8), allowing method optimization without compromising peak shape.