Learn More
Waters Corp Waters Column, SymmetryShield RP18, 5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186000217
Encompass_Preferred
The SymmetryShield RP18 Column is designed for analytical separations. It features a unique shielding technology that provides enhanced peak shape for basic compounds, making it suitable for various chromatographic applications.
  • 5 μm particle size
  • 2.1 mm x 50 mm column dimensions
  • Enhanced peak shape for basic compounds
  • Suitable for analytical separations
Catalog No. 50-780-136