Learn More
Waters Corp Waters Column, C18, 5 μm, 4.6 mm x 100 mm, 100 Å
Catalog No. 50781247 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns incorporate Waters’ patented embedded polar group technology, which “shields” the silica’s residual silanol surface from highly basic analytes. This reduced silanol activity significantly improves peak shape and resolution.
  • Utilizes patented embedded polar group technology to shield against basic analytes.
  • Provides improved peak shape and resolution across a broad pH range.
  • Retains basic chemicals less than conventional C18 and C8 columns.
  • Offers tight specification ranges for outstanding reproducibility, minimizing resolution shifts.
  • Produces more consistent results from batch to batch and column to column.
  • Long-lasting due to hydrolytic stability and mechanical stability of packed bed, lowering lab costs.
  • Provides a wide pH range (2 to 8 pH) for optimization at neutral or low pH.
Catalog No. 50-781-247 Supplier Waters Corp Supplier No. 186002618
Add to Cart
Add to Cart
These reversed-phase columns incorporate Waters’ patented embedded polar group technology, which “shields” the silica’s residual silanol surface from highly basic analytes. This reduced silanol activity significantly improves peak shape and resolution.
  • Utilizes patented embedded polar group technology to shield against basic analytes.
  • Provides improved peak shape and resolution across a broad pH range.
  • Retains basic chemicals less than conventional C18 and C8 columns.
  • Offers tight specification ranges for outstanding reproducibility, minimizing resolution shifts.
  • Produces more consistent results from batch to batch and column to column.
  • Long-lasting due to hydrolytic stability and mechanical stability of packed bed, lowering lab costs.
  • Provides a wide pH range (2 to 8 pH) for optimization at neutral or low pH.