Learn More
Waters Corp Waters Symmetry Shield RP18 Column, 100Å, 5 μm, 4.6 mm x 250 mm
Supplier:  Waters Corp 186000112
Encompass_Preferred
These reversed-phase columns offer high-performance separations with innovative embedded polar group technology. Designed to improve peak shape and resolution, these columns provide stable retention across a wide pH range and in highly aqueous mobile phases, ensuring reproducible results for demanding applications.
  • Embedded polar group technology shields silica from basic analytes, improving peak shape and resolution.
  • Achieves USPW Tailing Factors less than 1.2 over a pH range of 2 to 8.
  • Water-wettable particles ensure stable retention and superior peak shape in highly aqueous mobile phases.
  • Ensures reproducibility through high purity silica and tightly controlled manufacturing.
  • Each column includes a Certificate of Analysis and Performance Test Chromatogram.
Catalog No. 50-780-072