Learn More
Waters Corp Symmetry Shield RP18 Prep Column, 100Å, 7 µm, 19 mm X 150 mm, 1/pk
Catalog No. 50780769 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP18 column significantly improves peak shape and resolution.
Catalog No. 50-780-769 Supplier Waters Corp Supplier No. 186001839
Add to Cart
Add to Cart
Request a Quote
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP18 column significantly improves peak shape and resolution.