Learn More
Waters Corp Sentry Guard Cartridge, Symmetry Shield RP18, 3.5 μm, 3.9 mm x 20 mm
Catalog No. 50780467 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Features patented embedded polar group technology, shielding silica from highly basic analytes.
  • Reduces silanol activity for improved peak shape and resolution.
  • Extends analytical column lifespan; prevents pollutants from reaching the analytical column.
  • Utilizes high-performance packing materials, engineered for biological fluid cleanup and temperature control.
  • Manufactured in cGMP, ISO 9001-certified facility; chromatographically tested for consistent, repeatable performance.
  • Withstands up to 6000 psi (415 bar) of pressure and operates in a pH range of 2 to 8.
Catalog No. 50-780-467 Supplier Waters Corp Supplier No. 186000701
  • Features patented embedded polar group technology, shielding silica from highly basic analytes.
  • Reduces silanol activity for improved peak shape and resolution.
  • Extends analytical column lifespan; prevents pollutants from reaching the analytical column.
  • Utilizes high-performance packing materials, engineered for biological fluid cleanup and temperature control.
  • Manufactured in cGMP, ISO 9001-certified facility; chromatographically tested for consistent, repeatable performance.
  • Withstands up to 6000 psi (415 bar) of pressure and operates in a pH range of 2 to 8.