Learn More
Waters Corp Waters Sentry Guard Cartridge, C18, 100 Å, 5 μm
Supplier:  Waters Corp 186000107
Encompass_Preferred
These reversed-phase guard cartridges feature embedded polar group technology to “shield” the silica’s residual silanol surface from basic analytes, improving peak shape and resolution. They extend analytical column lifespan by limiting contaminating chemicals and suspended particles, and are made with high-performance packing materials for biological fluid purification and temperature control.
  • Embedded polar group technology “shields” silica’s residual silanol surface from basic analytes, improving peak shape and resolution.
  • Extends analytical column lifespan by limiting contaminating chemicals and suspended particles.
  • Made with high-performance packing materials.
  • Designed for biological fluid purification and/or temperature control.
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure chemicals, with chromatographic testing for each batch.
Catalog No. 50-780-067