Learn More
Waters Corp Waters Symmetry Shield RP8 Column, 100Å, 3.5 μm, 3 mm X 100 mm
Catalog No. 50780469 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns incorporate patented embedded polar group technology, which “shields” the residual silanol surface of the silica from highly basic analytes. This reduction in silanol activity significantly improves peak shape and resolution.
  • High repeatability for ongoing HPLC procedure compliance.
  • Embedded polar group technology protects silanols from basic analytes, enhancing peak form.
  • Close proximity of embedded polar group to silica surface provides distinct selectivity and retention.
  • Manufactured in a cGMP-certified, ISO 9001-certified facility using purest reagents.
Catalog No. 50-780-469 Supplier Waters Corp Supplier No. 186000703
These reversed-phase columns incorporate patented embedded polar group technology, which “shields” the residual silanol surface of the silica from highly basic analytes. This reduction in silanol activity significantly improves peak shape and resolution.
  • High repeatability for ongoing HPLC procedure compliance.
  • Embedded polar group technology protects silanols from basic analytes, enhancing peak form.
  • Close proximity of embedded polar group to silica surface provides distinct selectivity and retention.
  • Manufactured in a cGMP-certified, ISO 9001-certified facility using purest reagents.