Learn More
Waters Corp Symmetry Shield RP8 Column, 100Å, 3.5 µm, 2.1 mm X 50 mm, 1/pk
Catalog No. 50786002 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP8 column significantly improves peak shape and resolution.
Catalog No. 50-786-002 Supplier Waters Corp Supplier No. WAT094257
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP8 column significantly improves peak shape and resolution.