Learn More
Waters Corp Waters Column, Symmetry Shield RP8, 3.5 μm, 4.6 mm ID x 100 mm
Catalog No. 50786005 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns feature Waters’ patented embedded polar group technology, which “shields” the silica’s residual silanol surface from highly basic analytes. The SymmetryShield RP8 column significantly improves peak shape and resolution due to reduced silanol activity.
  • Contains patented embedded polar group technology that “shields” the silica’s residual silanol surface from highly basic analytes.
  • Reduced silanol activity significantly improves peak shape and resolution.
  • Provides a high degree of reproducibility and complete confidence in the long-term compliance of HPLC procedures.
  • The embedded polar group near the silica surface decreases the activity of surface silanols.
  • Imparts different selectivity and retention compared to the C18 ligand.
  • Manufactured in a cGMP, ISO 9001-certified facility with ultra-pure reagents.
Catalog No. 50-786-005 Supplier Waters Corp Supplier No. WAT094266
These reversed-phase columns feature Waters’ patented embedded polar group technology, which “shields” the silica’s residual silanol surface from highly basic analytes. The SymmetryShield RP8 column significantly improves peak shape and resolution due to reduced silanol activity.
  • Contains patented embedded polar group technology that “shields” the silica’s residual silanol surface from highly basic analytes.
  • Reduced silanol activity significantly improves peak shape and resolution.
  • Provides a high degree of reproducibility and complete confidence in the long-term compliance of HPLC procedures.
  • The embedded polar group near the silica surface decreases the activity of surface silanols.
  • Imparts different selectivity and retention compared to the C18 ligand.
  • Manufactured in a cGMP, ISO 9001-certified facility with ultra-pure reagents.