Learn More
Waters Corp Waters Column, Reversed Phase, C8, 3.5 μm, 100 Å, 3 mm x 150 mm
Catalog No. 50780468 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These reversed-phase columns feature patented embedded polar group technology that “shields” the residual silanol surface of the silica from highly basic analytes. The column’s reduced silanol activity significantly improves peak shape and resolution.
  • High reproducibility through narrow ligand surface coverage.
  • Improved peak shape and resolution via embedded polar group technology.
  • Reduced retention of basic compounds, aiding method development.
  • Enhanced peak form over 2–8 pH range.
  • Optimized for low organic/highly aqueous mobile phases.
  • Prevents pore dewetting, ensuring analyte retention.
  • Increased laboratory productivity through faster procedure development.
Catalog No. 50-780-468 Supplier Waters Corp Supplier No. 186000702
Add to Cart
Add to Cart
These reversed-phase columns feature patented embedded polar group technology that “shields” the residual silanol surface of the silica from highly basic analytes. The column’s reduced silanol activity significantly improves peak shape and resolution.
  • High reproducibility through narrow ligand surface coverage.
  • Improved peak shape and resolution via embedded polar group technology.
  • Reduced retention of basic compounds, aiding method development.
  • Enhanced peak form over 2–8 pH range.
  • Optimized for low organic/highly aqueous mobile phases.
  • Prevents pore dewetting, ensuring analyte retention.
  • Increased laboratory productivity through faster procedure development.