Learn More
Waters Corp Waters Symmetry Shield RP8 Column, 100Å, 5 μm, 4.6 mm × 250 mm
Supplier:  Waters Corp WAT200670
Encompass_Preferred
These reversed-phase columns, known as Symmetry Shield RP8 Columns, incorporate Waters’ innovative embedded polar group technology. This technology “shields” the residual silanol surface of the silica from highly basic analytes, significantly enhancing peak shape and resolution through reduced silanol activity.
  • Improved peak shape and resolution across a broad pH range of 2 to 8.
  • Less retention of basic compounds compared to conventional C18 and C8 columns.
  • Achieves USP Tailing Factors of less than 1.2.
  • Enables faster new method development and better separation.
  • Water-wettable particles ensure stable retention in highly aqueous mobile phases with superior peak shape.
  • Includes a Certificate of Analysis and a Performance Test Chromatogram.
  • Manufactured to stringent standards for excellent reproducibility.
Catalog No. 50-786-117