Learn More
Waters Corp Symmetry Shield RP8 Prep Column, 100Å, 5 µm, 19 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186001844
Encompass_Preferred
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP8 column significantly improves peak shape and resolution.
Catalog No. 50-780-774
Add to Cart
Request a Quote