Learn More
Waters Corp Symmetry Shield RP8 Prep Column, 100Å, 5 µm, 19 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186001842
Encompass_Preferred
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP8 column significantly improves peak shape and resolution.
Catalog No. 50-780-772