Learn More
Waters Corp Waters Symmetry Shield RP8 Column, 5 μm, 100Å, 2.1 x 150 mm
Catalog No. 50785998 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Symmetry Shield RP8 Column is engineered for high-performance liquid chromatography. With a 5 μm particle size, 100Å pore size, and 2.1 x 150 mm dimensions, it optimizes peak shape and resolution for basic compounds across various analytical applications.
  • Delivers improved peak shape and resolution over a broad pH range.
  • Exhibits less retention of basic compounds than conventional C18 and C8 columns.
  • Achieves USP Tailing Factors less than 1.2 over a pH range of 2 to 8.
  • Enables faster method development and improves laboratory productivity through lower detection limits.
  • Offers unique selectivity.
  • Ideal for highly aqueous mobile phases, providing stable retention and superior peak shape.
  • Prevents pore dewetting, ensuring consistent analyte retention without lengthy re-equilibration.
Catalog No. 50-785-998 Supplier Waters Corp Supplier No. WAT094245
The Waters Symmetry Shield RP8 Column is engineered for high-performance liquid chromatography. With a 5 μm particle size, 100Å pore size, and 2.1 x 150 mm dimensions, it optimizes peak shape and resolution for basic compounds across various analytical applications.
  • Delivers improved peak shape and resolution over a broad pH range.
  • Exhibits less retention of basic compounds than conventional C18 and C8 columns.
  • Achieves USP Tailing Factors less than 1.2 over a pH range of 2 to 8.
  • Enables faster method development and improves laboratory productivity through lower detection limits.
  • Offers unique selectivity.
  • Ideal for highly aqueous mobile phases, providing stable retention and superior peak shape.
  • Prevents pore dewetting, ensuring consistent analyte retention without lengthy re-equilibration.