Learn More
Aviva Systems Biology Corp SYMPK Antibody - N-terminal region (ARP42900_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42900_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYMPK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SYMPK. Alternate names: FLJ27092, SPK, SYMApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58355