Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

syn-Benzaldehyde Oxime 99+%, Thermo Scientific™

Catalog No. AC278911000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC278911000 100 g
AC278910250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC278911000 Supplier Thermo Scientific Chemicals Supplier No. 278911000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-90-1
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00002119 MFCD00063017
InChI Key VTWKXBJHBHYJBI-SOFGYWHQSA-N
Synonym benzaldehyde oxime, benzaldoxime, benzaldehyde, oxime, e-benzaldehyde oxime, alpha-benzaldoxime, benzaldehydeoxime, syn-benzaldoxime, benzaldehyde, oxime, e, e-n-phenylmethylidene hydroxylamine, z-benzaldoxime
PubChem CID 5324611
IUPAC Name (E)-N-(phenylmethylidene)hydroxylamine
SMILES O\N=C\C1=CC=CC=C1

Specifications

Color Yellow
Infrared Spectrum Authentic
Linear Formula C6H5CH=NOH
Refractive Index 1.5900 to 1.5920 (20°C, 589nm)
Quantity 100 g
Formula Weight 121.14
Percent Purity ≥99.0%
Physical Form Liquid After Melting
Chemical Name or Material syn-Benzaldehyde oxime, 99+%
EINECSNumber 213-261-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.