Learn More
Medchemexpress LLC HY-18307 5mg Medchemexpress, SYN1143 CAS:913376-84-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY183075MG
Encompass_Preferred
Medchemexpress, HY-18307 5mg SYN1143 CAS:913376-84-8 SYN1143 is a potent, selective and orally active dual inhibitor of c-Met/RON, with IC50s of 4 and 9 nM, respectively. SYN1143 has weak inhibitory activity on Lck, Tie2, Src, and BTK with IC50s ranging from 160 to 710 nM. SYN1143 can be used for the research of cancers that RON and c-Met are activated. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2036