Learn More
CPC Scientific 5TAMRA-Leu-Arg-Arg-Arg-Leu-Ser-Asp-Ala-Asn-Phe-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PROK005A
Encompass
TAMRA-labeled Synapsin I derived peptide (Ab/Em = 544/574 nm) that is a neuron specific substrate for cAMP dependent and Ca2+/calmodulin dependent protein kinase. It plays an important role in the mechanism involved in neuron transmitter disease and also regulates the availability of synaptic vesicles for exocytosis. ONE-LETTER SEQUENCE: 5TAMRA-LRRRLSDANF-NH2MOLECULAR FORMULA: C78H111N23O18MOLECULAR WEIGHT:1658.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Bahler, M. et al. J. Neurochem. 73, 921 (1999)
Catalog No. 50-211-0225