Learn More
Waters Corp SYNAPT G2 NanoLockSpray Performance Maintenance Kit
Catalog No. 50466525 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit for the Synapt G2 Mass Spectrometer with NanoLockSpray Source option contains all the Waters Quality Parts necessary to keep your instrument operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts: Source component, ESI probe components, and vacuum pump components.
Catalog No. 50-466-525 Supplier Waters Corp Supplier No. 201000230
Add to Cart
Add to Cart
Request a Quote
The Waters Performance Maintenance Kit for the Synapt G2 Mass Spectrometer with NanoLockSpray Source option contains all the Waters Quality Parts necessary to keep your instrument operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts: Source component, ESI probe components, and vacuum pump components.