Learn More
Waters Corp Performance Maintenance Kit, SYNAPT G2-S, LockSpray (Scroll), Mass Spectrometer
Catalog No. 502391228 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the Synapt G2-S (Lockspray / scroll) Mass Spectrometer contains all the necessary parts to keep your instrument operating at peak performance on an annual basis.
  • Contains all the necessary parts to keep your instrument operating at peak performance on an annual basis.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Designed for the Synapt G2-S (Lockspray / scroll) Mass Spectrometer.
Catalog No. 50-239-1228 Supplier Waters Corp Supplier No. 201000255
The Performance Maintenance Kit for the Synapt G2-S (Lockspray / scroll) Mass Spectrometer contains all the necessary parts to keep your instrument operating at peak performance on an annual basis.
  • Contains all the necessary parts to keep your instrument operating at peak performance on an annual basis.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Designed for the Synapt G2-S (Lockspray / scroll) Mass Spectrometer.