Learn More
Waters Corp Performance Maintenance Kit, SYNAPT G2-Si LockSpray (Rotary), with Chemical Kit
Catalog No. 502392159 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides all the necessary quality parts to maintain the Synapt G2-Si (LockSpray / rotary) Mass Spectrometer annually, ensuring peak performance. It includes source components, ESI probe components, vacuum pump components, and a chemical kit required for performance testing.
  • Designed for performance maintenance.
  • For MS system type.
  • Recommended for annual replacement.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Includes a chemical kit for performance testing.
Catalog No. 50-239-2159 Supplier Waters Corp Supplier No. 176003674
This kit provides all the necessary quality parts to maintain the Synapt G2-Si (LockSpray / rotary) Mass Spectrometer annually, ensuring peak performance. It includes source components, ESI probe components, vacuum pump components, and a chemical kit required for performance testing.
  • Designed for performance maintenance.
  • For MS system type.
  • Recommended for annual replacement.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Includes a chemical kit for performance testing.