Learn More
Waters Corp Waters Corporation O-Ring, FKM Fluoroelastomer, 10 mm ID, 1.5 mm CS
Catalog No. NC9017927 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement O-ring is constructed from FKM Fluoroelastomer and features precise dimensions of 10 mm internal diameter by 1.5 mm cross-section. It functions as a critical seal between the sample cone and the ion block in mass spectrometers, specifically those incorporating the Universal Source technology, like the Xevo product range.
  • Made of FKM Fluoroelastomer material.
  • Internal diameter of 10 mm.
  • Thickness of 1.5 mm.
  • Designed as a seal for mass spectrometers.
  • Typically used between the sample cone and the ion block.
  • Compatible with Universal Source mass spectrometers.
  • Packaged as a single unit.
  • Replaced as required.
Catalog No. NC9017927 Supplier Waters Corp Supplier No. 700004321
This replacement O-ring is constructed from FKM Fluoroelastomer and features precise dimensions of 10 mm internal diameter by 1.5 mm cross-section. It functions as a critical seal between the sample cone and the ion block in mass spectrometers, specifically those incorporating the Universal Source technology, like the Xevo product range.
  • Made of FKM Fluoroelastomer material.
  • Internal diameter of 10 mm.
  • Thickness of 1.5 mm.
  • Designed as a seal for mass spectrometers.
  • Typically used between the sample cone and the ion block.
  • Compatible with Universal Source mass spectrometers.
  • Packaged as a single unit.
  • Replaced as required.