Learn More
Waters Corp Performance Maintenance Kit, SYNAPT XS, MS System, Annual
Catalog No. 502391457 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This performance maintenance kit contains all the necessary quality parts to keep your SYNAPT XS Mass Spectrometer operating at peak performance on an annual basis.
  • Kit contains source components.
  • Kit contains air filters.
  • Kit contains a lock spray reference probe.
  • A probe assembly is not included, but it is highly recommended to order or replace the appropriate probe assembly during performance maintenance for peak performance.
Catalog No. 50-239-1457 Supplier Waters Corp Supplier No. 201000326
This performance maintenance kit contains all the necessary quality parts to keep your SYNAPT XS Mass Spectrometer operating at peak performance on an annual basis.
  • Kit contains source components.
  • Kit contains air filters.
  • Kit contains a lock spray reference probe.
  • A probe assembly is not included, but it is highly recommended to order or replace the appropriate probe assembly during performance maintenance for peak performance.