Learn More
Cell Signaling Technology Synaptic Neuron Marker Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502731520
Encompass
The Synaptic Neuron Marker Antibody Sampler Kit provides an economical means of detecting presynaptic and postsynaptic proteins by western blot. Each kit contains a selection of primary antibodies validated for studying synaptic development and protein markers under normal and disease conditions. These antibodies are supplied in a storage buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Includes primary antibodies to perform at least two western blot experiments with each antibody.
  • Enables detection of both presynaptic and postsynaptic protein markers.
  • Features rabbit monoclonal antibodies for high specificity and performance.
  • Suitable for applications including western blotting, immunoprecipitation, and immunohistochemistry.
  • Supplied with an HRP-linked secondary antibody for convenient detection.
Catalog No. 50-273-1520 Supplier Cell Signaling Technology Supplier No. 74798T
The Synaptic Neuron Marker Antibody Sampler Kit provides an economical means of detecting presynaptic and postsynaptic proteins by western blot. Each kit contains a selection of primary antibodies validated for studying synaptic development and protein markers under normal and disease conditions. These antibodies are supplied in a storage buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Includes primary antibodies to perform at least two western blot experiments with each antibody.
  • Enables detection of both presynaptic and postsynaptic protein markers.
  • Features rabbit monoclonal antibodies for high specificity and performance.
  • Suitable for applications including western blotting, immunoprecipitation, and immunohistochemistry.
  • Supplied with an HRP-linked secondary antibody for convenient detection.