Learn More
Biotium SynaptoGreen™C2
Catalog No. 501964761
Encompass_Preferred
SynaptoGreen™ C2 is more water soluble than SynaptoGreen™ C4 and thus has a faster de-staining rate than the latter. Properties: Red solid soluble in water; C26H41Br2N3; MW: 556.FM2-10 is a registered trademark of Thermo Fisher Scientific.
Catalog No. 50-196-4761 Supplier Biotium Supplier No. 70044
Add to Cart
Add to Cart
SynaptoGreen™ C2 is more water soluble than SynaptoGreen™ C4 and thus has a faster de-staining rate than the latter. Properties: Red solid soluble in water; C26H41Br2N3; MW: 556.FM2-10 is a registered trademark of Thermo Fisher Scientific.