Learn More
Biotium SynaptoGreen™C2 5x1mg
Supplier:  Biotium 70045
Encompass_Preferred
SynaptoGreen™ C2 is more water soluble than SynaptoGreen™ C4 and thus has a faster de-staining rate than the latter. Properties: Red solid soluble in water; C26H41Br2N3; MW: 556.FM2-10 is a registered trademark of Thermo Fisher Scientific.
Catalog No. 50-196-4762