Learn More
Biotium SynaptoGreen™ C4, 5MG (Equivalent to FM1-43)
Catalog No. NC1669443
Encompass_Preferred
A widely used green fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. Biotium offers several Nerve Terminal Staining Kits for staining brain slices with synaptic vesicle dyes.Features: Ex/Em nm: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C30H49Br2N3; MW: 612 FM is a registered trademark of Thermo Fisher Scientific.
Catalog No. NC1669443 Supplier Biotium Supplier No. 70020
A widely used green fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. Biotium offers several Nerve Terminal Staining Kits for staining brain slices with synaptic vesicle dyes.Features: Ex/Em nm: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C30H49Br2N3; MW: 612 FM is a registered trademark of Thermo Fisher Scientific.