Learn More
Biotium SynaptoGreen™C4 5x1mg
Catalog No. 501964752
Encompass_Preferred
A widely used green fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. Biotium offers several Nerve Terminal Staining Kits for staining brain slices with synaptic vesicle dyes.Properties: Ex/Em: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C30H49Br2N3; MW: 612.FM is a registered trademark of Thermo Fisher Scientific.
Catalog No. 50-196-4752 Supplier Biotium Supplier No. 70022
A widely used green fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. Biotium offers several Nerve Terminal Staining Kits for staining brain slices with synaptic vesicle dyes.Properties: Ex/Em: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C30H49Br2N3; MW: 612.FM is a registered trademark of Thermo Fisher Scientific.