Learn More
Diagnostic Biosystems SYNAPTOPHYSIN SYP02 Synaptophysin (SYP02)
Catalog No. 502407322
Encompass_Preferred
This antibody isspecific to a 38 kDa protein. Synaptophysin is an glycoprotein occurring in presynaptic vesicles of neurons in the brain, spinal cord, retina, vesicles of adrenal medulla and in neuromuscular junctions. This antibody reacts with neuroendocrine neoplasms of neural as well as epithelial types.
Catalog No. 50-240-7322 Supplier Diagnostic Biosystems Supplier No. PDM592
This antibody isspecific to a 38 kDa protein. Synaptophysin is an glycoprotein occurring in presynaptic vesicles of neurons in the brain, spinal cord, retina, vesicles of adrenal medulla and in neuromuscular junctions. This antibody reacts with neuroendocrine neoplasms of neural as well as epithelial types.