Learn More
Cayman Chemical SynaptophysInCTRm RabbIt 100uL
SDP
Catalog No. 502949168
Encompass_Preferred
A DGK inhibitor (Ki 58 M) reduces basal and thrombin-stimulated phosphatidic acid levels in washed isolated human platelets at 30 100 and 500 M
Catalog No. 50-294-9168 Supplier Cayman Chemical Supplier No. 32213100 UL
A DGK inhibitor (Ki 58 M) reduces basal and thrombin-stimulated phosphatidic acid levels in washed isolated human platelets at 30 100 and 500 M