Learn More
Biotium SynaptoRed™C1
Catalog No. 501964760
Encompass_Preferred
SynaptoRed™ C1 is similar to the widely used red fluorescent nerve terminal dye SynaptoRed™ C2 (also called FM4-64) except that the lipophilic tail of SynaptoRed™ C1 is one carbon shorter than that of the SynaptoRed™ C2. Thus, SynaptoRed™ C1 is expected to have a faster off-rate compared to SynaptoRed™ C2. Properties: Dark purple solid soluble in water; C28H41Br2N3; MW: 580
Catalog No. 50-196-4760 Supplier Biotium Supplier No. 70040
SynaptoRed™ C1 is similar to the widely used red fluorescent nerve terminal dye SynaptoRed™ C2 (also called FM4-64) except that the lipophilic tail of SynaptoRed™ C1 is one carbon shorter than that of the SynaptoRed™ C2. Thus, SynaptoRed™ C1 is expected to have a faster off-rate compared to SynaptoRed™ C2. Properties: Dark purple solid soluble in water; C28H41Br2N3; MW: 580