Learn More
Biotium Synaptored™C2 (also known as FM4-64)
Catalog No. NC1679289
Encompass_Preferred
A widely used red fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. When used in combination with the green fluorescent dye SynaptoGreen™, synapses or neuromuscular junctions can be imaged independently in two colors. SynaptoRed™ has also been used to study endocytosis in Dictyostelia.Features: Dark purple solid soluble in water; C30H45Br2N3; MW: 608; [162112-35-8] FM is a registered trademark of Thermo Fisher Scientific.
Catalog No. NC1679289 Supplier Biotium Supplier No. 70021
A widely used red fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. When used in combination with the green fluorescent dye SynaptoGreen™, synapses or neuromuscular junctions can be imaged independently in two colors. SynaptoRed™ has also been used to study endocytosis in Dictyostelia.Features: Dark purple solid soluble in water; C30H45Br2N3; MW: 608; [162112-35-8] FM is a registered trademark of Thermo Fisher Scientific.