Learn More
Biotium SynaptoRed™C2 5x1mg
Catalog No. 501964755
Encompass_Preferred
A widely used red fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. When used in combination with the green fluorescent dye SynaptoGreen™, synapses or neuromuscular junctions can be imaged independently in two colors. SynaptoRed™ has also been used to study endocytosis in Dictyostelia. Properties: Dark purple solid soluble in water C30H45Br2N3; MW: 608; [162112-35-8].FM is a registered trademark of Thermo Fisher Scientific.
Catalog No. 50-196-4755 Supplier Biotium Supplier No. 70027
A widely used red fluorescent dye for following synaptic activity by staining synaptic vesicles at the synapse or neuromuscular junctions. When used in combination with the green fluorescent dye SynaptoGreen™, synapses or neuromuscular junctions can be imaged independently in two colors. SynaptoRed™ has also been used to study endocytosis in Dictyostelia. Properties: Dark purple solid soluble in water C30H45Br2N3; MW: 608; [162112-35-8].FM is a registered trademark of Thermo Fisher Scientific.