Learn More
Biotium SynaptoRed™C2M
Catalog No. 501964756
Encompass_Preferred
SynaptoRed™ C2M is a red fluorescent probe used to follow synaptic vesicles. SynaptoRed™ C2M is slightly more water soluble than SynaptoRed™ C2 (equivalent to FM4-64). FM is a registered trademark of Thermo Fisher Scientific.
Catalog No. 50-196-4756 Supplier Biotium Supplier No. 70028
SynaptoRed™ C2M is a red fluorescent probe used to follow synaptic vesicles. SynaptoRed™ C2M is slightly more water soluble than SynaptoRed™ C2 (equivalent to FM4-64). FM is a registered trademark of Thermo Fisher Scientific.