Learn More
Enzo Life Sciences Synaptotagmin monoclonal antibody (ASV30) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISYA130F
Encompass_Preferred
Clone: ASV30. Host: Mouse. Isotype: IgG2a. Immunogen: Rat brain synaptic junction protein complexes. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7678