Learn More
Synoptics™ Synbiosis Bar Code Reader, CCD Sensor
GREENER_CHOICESDP
Catalog No. 01000812 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Display Type:
None
Catalog No. Display Type
01-000-812 None
1 options
Catalog No. 01-000-812 Supplier Synoptics™ Supplier No. BARCO
Add to Cart
Add to Cart
ProtoCOL Bar Code Reader is designed for instant reading of plate information on Proc3, Protos 3, ChromaZona, and aCOL3, with a CCD sensor type offering a wide reader area.
  • Handheld, USB, barcode scanner to enable instant reading of plate information
  • CCD type sensor provides wide reading area
  • Suitable for use with all ProtoCOL 3 systems
Please note: for the barcode reader to function within the ProtoCOL 3 software, the ProtoCOL 3 media key or licensing dongle must be modified. If the barcode reader is bought in conjunction with a ProtoCOL 3 system, then this will be done automatically. If the barcode reader is bought separately, for use with an existing ProtoCOL 3 system, then the media key or licensing dongle pertaining to that system must be returned to us so that we can enable the barcode reading features in the software.
Specifications
Product Type Barcode Reader
Display Type None
Inlet Style None
Magnification None
Material Polycarbonate
Memory None
Outlet Style USB
Sample Size Single
For Use With (Application) Reading Barcodes on Plates
For Use With (Equipment) Proc3, Protos 3, aCOL3, and ChromaZona
WARNING: Cancer - www.P65Warnings.ca.gov