Learn More
Synoptics™ Synbiosis Validation Kit
GREENER_CHOICESDP
Catalog No. 01000806 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Validation of Aone Imaging
Validation of Color Count Imaging
Validation of Count Plate Imaging
Validation of East Imaging
Validation of Opka Imaging
Validation of Pour Plate Imaging
Validation of SBA Imaging
Validation of Spiral Imaging
For Use With (Equipment):
ProtoCOL
ProtoCOL and ChromaZona
ProtoCOL and Protos
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) For Use With (Equipment)
01-000-806 Validation of Color Count Imaging ProtoCOL and Protos
01-000-804 Validation of Pour Plate Imaging ProtoCOL and Protos
01-000-805 Validation of Count Plate Imaging ProtoCOL and Protos
01-000-809 Validation of Spiral Imaging ProtoCOL and Protos
01-000-807 Validation of Aone Imaging ProtoCOL and ChromaZona
01-000-808 Validation of East Imaging ProtoCOL and ChromaZona
01-000-810 Validation of Opka Imaging ProtoCOL
01-000-811 Validation of SBA Imaging ProtoCOL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-000-806 Supplier Synoptics™ Supplier No. SYNBKITCOLOURCLASS
Add to Cart
Add to Cart
Synbiosis Validation Kits are designed for use with ProtoCOL, Protos, and ChromaZona instruments and include a validation plate, a validation procedure, and a test certificate.
  • Synbiosis Validation Kits are designed for use with ProtoCOL, Protos, and ChromaZona instruments
  • Each kit includes a validation plate, a validation procedure, and a test certificate
  • Kits available for validation of: Pour Plate Imaging, Count Plate Imaging, Color Count Imaging, Aone Imagine, East Imaging, Spiral Imaging, Opka Imaging, SBA Imaging
Specifications
Product Type Validation Kit
Display Type None
Inlet Style None
Magnification None
Material Plastic, Paper
Memory None
Outlet Style None
Sample Size Single
For Use With (Application) Validation of Color Count Imaging
For Use With (Equipment) ProtoCOL and Protos
Includes RBG Color Validation Plate, Color Classification Validation Procedure, Color Test Certificate