Learn More
Aviva Systems Biology Corp SYNC Antibody - C-terminal region (ARP63616_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63616_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYNC. It was validated on Western Blot. . Alternate names: MGC149625, MGC149626, SYNC1, SYNCOILINApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-032